missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RbAp46 (aa 1-13) Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Human RbAp46 (residues 1-13) Synthetic Peptide, PEP-083, from Invitrogen.
PEP-083 is a 13 amino acid synthetic peptide whose sequence corresponds to residues 1-13 of human RbAp46 with a carboxy-terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): M1-A-S-K-E-M-F-E-D-T-V-E-E13-CThis peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and endogenous RbAp46, catalog # PA1-868 and PA1.1-868. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
Specifications
Specifications
| Gene ID (Entrez) | 5931 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q16576 |
| Common Name | Human RbAp46 (aa 1-13) |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Lyophilized |
| Formulation | peptide with no preservative |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction