missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RBM5 (aa 343-472) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53586 (PA5-53586. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
DEF-3 and LUCA15 belong to an evolutionarily conserved family of RNA binding proteins and share similar expression patterns. Both DEF-3 and LUCA15 are highly expressed in adult heart and thymus as well as fetal kidney. Conversely, fetal thymus and adult kidney express very little DEF-3 and LUCA15. In the haemopoietic system of mice, the expression of DEF-3 is downregulated upon differentiation of progenitor cells into granulocytes but persists during macrophage development. Both DEF-3 and LUCA15 contain two zinc finger motifs, a bipartite nuclear signal and two RNA binding motifs. DEF-3 and LUCA15 are capable of specifically binding poly(G) RNA.
Spezifikation
Spezifikation
| Accession Number | P52756 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 10181 |
| Name | Human RBM5 (aa 343-472) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | D030069N10Rik; G15; H37; LUCA15; Protein G15; Putative tumor suppressor LUCA15; Rbm5; Renal carcinoma antigen NY-REN-9; RMB5; RNA binding motif protein 5; RNA-binding motif protein 5; RNA-binding protein 5 |
| Common Name | RBM5 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur