missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant BMP-2

Product Code. 17129321
Encompass_Preferred
Click to view available options
Quantity:
100 μg
1000 μg
50 μg
Unit Size:
1000µg
100µg
50µg
This item is not returnable. View return policy

Product Code. 17129321

Brand: STEMCELL Technologies 78004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bone morphogenetic protein 2

  • Bone morphogenetic protein 2 (BMP-2) is a member of the transforming growth factor beta (TGF-β) superfamily
  • BMP-2 is a disulfide-linked homodimer, acts as a ligand for complexes of type I and II BMP receptors, and primarily activates SMAD1/5/8 signaling (Nohe et al.) BMP-2 is a potent differentiation factor and directs human pluripotent stem cells (hPSCs) towards various cell types including extra-embryonic endoderm, mesenchymal cells, and chondrocytes (Pera et al.) Although BMP-2 expression is low in healthy cartilage, its expression is upregulated at the site of cartilage damage (Blaney Davidson et al.) BMP-2 induces bone and cartilage formation in vitro and is able to induce chondrogenesis in human mesenchymal stem cells (Schmitt et al.)

Specifications

Name BDA2, BMP-2A, BMP2, Bone morphogenetic protein 2A
Quantity 50 μg
Cell Type Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Pluripotent Stem Cells
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.