missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant Noggin

Product Code. 15888637
Encompass_Preferred
Click to view available options
Quantity:
10 μg
1000 μg
50 μg
Unit Size:
1000µg
10µg
50µg
This item is not returnable. View return policy

Product Code. 15888637

Brand: STEMCELL Technologies 78060

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Noggin

  • Noggin binds to and antagonizes bone morphogenetic protein (BMP) ligands of the transforming growth factor beta (TGF-β) family
  • Noggin supports maintenance of undifferentiated human embryonic stem cells in vitro, and can be used to prevent spontaneous differentiation in the short term (Chaturvedi et al.) Noggin is essential for development of structures derived from ectoderm embryonic somite, skeletal patterning, and neurogenesis in vivo
  • It also influences chondrogenesis, osteogenesis, and joint formation (Krause et al.), and promotes dopaminergic differentiation of embryonic stem cells and subsequent survival of dopamine neurons (Chiba et al.)

Specifications

Name NOG, SYM1, SYNS1, Symphalangism 1 (proximal), Synostoses (multiple) syndrome
Quantity 50 μg
Cell Type Neural Cells, PSC-Derived; Neurons; Pluripotent Stem Cells
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.