missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Human Resistin ELISA Kit

Product Code. 16861566 Shop All Proteintech Products
Change view
Click to view available options
Target Species:
Human
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Target Species unitSize
16861566 Human 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16861566 Supplier Proteintech™ Supplier No. KE0015996T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KE00159 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The Resistin ELISA kit is to be used to detect and quantify protein levels of endogenous Resistin.

  • The assay recognizes human Resistin. An antibody specific for Resistin has been pre-coated onto the microwells. The Resistin protein in samples is captured by the coated antibody after incubation.
  • Following extensive washing, another antibody specific for Resistin is added to detect the captured Resistin protein. For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent.
  • Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450nm with the correction wavelength set at 630 nm.
  • Resistin, a cysteine-rich hormone secreted primarily by rodent fat cells, was postulated to be implicated in obesity, insulin resistance and diabetes.
  • Resistin is also secreted by macrophages in humans, promoting inflammation (PMID: 20148674).
  • Resistin modulates the synthesis and secretion of key pro-inflammatory cytokines such as TNF-α, IL-6 and IL12 in macrophages via a nuclear factor-kB (NF-kB)-dependent pathway.
  • Given the connection between inflammation and cancer, resistin was soon considered for its role in cancer.
  • There were observations supporting its possible use as a diagnostic marker, expression levels of which also seemed to differ in patients with varying clinical stages (PMID: 30317640).
  • Resistin has been also linked to other pathologies such as atherosclerosis (PMID: 15710760).

Specifications

Accession Number BC069302
Assay Range 31.25 to 2000 pg/mL
Assay Sensitivity 15.2 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Human
Gene ID (Entrez) 56729
Host Species Human
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 4.5 hr.
UniProt ID Q9HD89
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human Resistin ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.