missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RREB1 (aa 1106-1196) Control Fragment Recombinant Protein

Product Code. 30201468
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201468

Brand: Invitrogen™ RP105888

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (77%), Rat (77%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65129 (PA5-65129. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a zinc finger transcription factor that binds to RAS-responsive elements (RREs) of gene promoters. It has been shown that the calcitonin gene promoter contains an RRE and that the encoded protein binds there and increases expression of calcitonin, which may be involved in Ras/Raf-mediated cell differentiation. Multiple transcript variants encoding several different isoforms have been found for this gene. [provided by RefSeq, Dec 2009].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q92766
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6239
Name Human RREB1 (aa 1106-1196) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1110037N09Rik; AA414966; B930013M22Rik; DNA-binding protein; FINB; Finger protein in nuclear bodies; hindsight homolog; HNT; LZ321; raf-responsive zinc finger protein LZ321; ras responsive element binding protein 1; ras-responsive element-binding protein 1; RAS-responsive zinc finger transcription factor RREB; RREB 1; RREB1; RREB-1; Zep 1; Zep1; Zep-1; zinc finger motif enhancer binding protein 1; zinc finger motif enhancer-binding protein 1; zinc finger motif-enhancer binding-protein 1
Common Name RREB1
Gene Symbol RREB1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GSVPKAATTATPAATTSPKESSEPPAPASSPEAASPTEQGPAGTSKKRGRKRGMRSRPRANSGGVDLDSSGEFASIEKMLATTDTNKFSPF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.