missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SCUBE3 Synthetic Peptide

Product Code. 15245379
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 15245379

Brand: Invitrogen™ PEP1469

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

SCUBE3 Synthetic Peptide, PEP-1469, from Invitrogen.

SCUBE3 is a member of a family of secreted glycoproteins that contain N-terminal EGF-like repeats and C-terminal cysteine-rich motifs and CUB domain and is highly expressed in primary osteoblasts and bones, and to a lesser extent in heart. Other studies have shown that overexpression of SCUBE3 in mice induced cardiac hypertrophy, suggesting that it may also play a role in the regulation of cardiac growth. SCUBE3 has been shown to be an endogenous TGF-Beta receptor ligand and is thought to promote lung cancer cell mobility and invasiveness. In lung cancer cells, the secreted SCUBE3 protein was cleaved by MMP2 and MMP9, allowing the activation of the TGF-Beta receptor, the increase of Smad2/3 transcriptional activity and the upregulation of expression of proteins such as TGF-Beta1, VEGF, Snail, and Slug.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 222663
Species Human
Purification Method purified
Accession Number Q8IX30
Common Name Human SCUBE3
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol SCUBE3
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.