missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Serpin A7/TBG DuoSet ELISA

Product Code. 18788327
Change view
Click to view available options
Quantity:
5 Plates
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18788327 5 Plates Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18788327 Supplier R&D Systems™ Supplier No. DY817605

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sufficient for at least five 96-well plates

The Human Serpin A7/TBG DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Serpin A7/TBG.
TRUSTED_SUSTAINABILITY

Specifications

Detectable Analytes Serpin A7/TBG
Assay ELISA Development Kit
Conjugate Biotin
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well microplate (sold separately)
For Use With (Application) ELISA
Gene ID (Entrez) 6906
Host Species Human
Measuring Range 78.1-5,000pg/mL
Quantity 5 Plates
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.