missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Serpin C1 Native Protein

Product Code. 11584161
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11584161 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11584161 Supplier Invitrogen™ Supplier No. RP43136
  • ONLINE EXCLUSIVE PRICE
    361.00 €
    321.29 € / 1mg
    Save 39.71 €
    11% Off
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Native Protein

RP-43136 contains native human Antithrombin III protein. RP-43136 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests.

The protein encoded by this gene is a plasma protease inhibitor and a member of the serpin superfamily. This protein inhibits thrombin as well as other activated serine proteases of the coagulation system, and it regulates the blood coagulation cascade. The protein includes two functional domains: the heparin binding-domain at the N-terminus of the mature protein, and the reactive site domain at the C-terminus. The inhibitory activity is enhanced by the presence of heparin. More than 120 mutations have been identified for this gene, many of which are known to cause antithrombin-III deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01008
Concentration 10.7 mg/mL
For Use With (Application) Control
Formulation 50% water with 50% glycerol and no preservative
Gene ID (Entrez) 462
Molecular Weight (g/mol) 58 kDa
Name Human Serpin C1
Quantity 1 mg
Source Human
Gene Alias AI114908; antithrombin; Antithrombin 3; anti-thrombin 3; antithrombin III; antithrombin-III; AT3; At-3; AT3D; ATIII; hypothetical protein LOC540261; MGC22579; PRO0309; serine (or cysteine) peptidase inhibitor, clade C (antithrombin), member 1; serine (or cysteine) proteinase inhibitor, clade C (antithrombin), member 1; serpin C1; serpin family C member 1; serpin peptidase inhibitor clade C member 1; serpin peptidase inhibitor, clade C (antithrombin), member 1; serpin peptidase inhibitor, clade C, member 1; Serpinc1; THPH7
Common Name SERPINC1
Gene Symbol SERPINC1
Conjugate Unconjugated
Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.