missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Siglec-2/CD22 DuoSet ELISA
Click to view available options
Sufficient For:
Five 96-well plates
Unit Size:
1 set
Description
The Human Siglec-2/CD22 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Siglec-2/CD22.
Specifications
Specifications
| Assay Range | 93.80 to 6,000pg/mL |
| Immunoassay Kit Format | 96-well Plate |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum |
| Sufficient For | Five 96-well plates |
| Target Species | Human |
| Gene ID (Entrez) | 933 (Human); 12483 (Mouse) |
| Kit Contents | 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Target | Sialic Acid Binding Ig-like Lectin 2 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction