missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Skeletal Muscle Membrane Tissue Lysate (Adult Membrane Diabetes)

Product Code. 18293530
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18293530 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18293530 Supplier Novus Biologicals™ Supplier No. NBL126450
  • ONLINE EXCLUSIVE PRICE
    768.00 €
    725.55 € / 0.1mg
    Save 42.45 €
    5% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The Human Skeletal Muscle Membrane Tissue Lysate (Adult Membrane Diabetes) is derived from human.
TRUSTED_SUSTAINABILITY

Specifications

Tissue Skeletal Muscle
Quantity 0.1 mg
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Membrane Protein - Diabetic Disease: Skeletal Muscle
Cellular Fraction Membrane
Tissue Condition Diabetes
Cross Reactivity Human
Host Species Human
Product Type Human Skeletal Muscle Membrane Tissue Lysate (Adult Diabetes)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.