missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLAIN2 Synthetic Peptide

Product Code. 15235369
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15235369 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15235369 Supplier Invitrogen™ Supplier No. PEP1458
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Synthetic Peptide

This peptide corresponds to 14 amino acid peptide near the amino terminus of human SLAIN2. PEP-1458 can be used as a blocking peptide with polyclonal antibody PA5-34416.

SLAIN2 is closely related to SLAIN1, a novel stem cell protein that is expressed at the stem cell and epiblast stages. Unlike SLAIN1, SLAIN2 is widely expressed and is thought to play a role in regulating microtubule (MT) growth and organization. SLAIN2 binds to end-binding proteins (EBs), cytoplasmic linker proteins (CLIPs) such as CLIP170, CLIP-associated proteins, and interacts with the MT polymerase ch-TOG. Depletion of SLAIN2 using RNAi led to disorganization of the radial MT array, indicating its importance in the regulation of MT growth and organization.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 57606
Species Human
Purification Method purified
Accession Number Q9P270
Common Name SLAIN2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 5033405K12Rik; 8030444K12Rik; AI596370; FLJ21611; hypothetical protein LOC534203; KIAA1458; LOW QUALITY PROTEIN: SLAIN motif-containing protein 2; RGD1311593; SLAIN motif family member 2; SLAIN motif family, member 2; SLAIN motif-containing protein 2; SLAIN2
Gene Symbol SLAIN2
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.