missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human SLITRK4 (NP_775101) VersaClone cDNA

Produktkod. 18748668
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 μg
Förpackningsstorlek
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
18748668 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18748668 missing translation for 'mfr' R&D Systems™ missing translation for 'supplierNo' RDC1779
  • ONLINE EXCLUSIVE PRICE
    791.00 €
    747.60 € / 10µg
    Save 43.40 €
    5% Off
 Restricted Item
Estimated Shipment: 08-09-2026
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy

SLIT and NTRK-like Family, Member 4

The Human SLITRK4 (NP_775101) VersaClone cDNA clone contains the complete ORF for SLITRK4, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The SLITRK4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Beställningsinfo

Ships at ambient temperature

TRUSTED_SUSTAINABILITY

Specifikationer

Product Type cDNA
Gene hSLITRK4
Accession Number NP_775101
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 2527bp
Gene ID (Entrez) 139065 (Human), 245446 (Mouse)
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.