missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SMO Partial ORF (NP_005622, 56 a.a. - 155 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006608-Q01.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16044735

  • 349.00 € / 10µg
Estimated Shipment: 29-06-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: GPPPPLSHCGRAAPCEPLRYNVCLGSVLPYGATSTLLAGDSDSQEEAHGKLVLWSGLRNAPRCWAVIQPLLCAVYMPKCENDRVELPSRTLCQATRGPCA
Specifications

Specifications

NP_005622
Liquid
6608
SMO (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gx/SMOH
SMO
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GPPPPLSHCGRAAPCEPLRYNVCLGSVLPYGATSTLLAGDSDSQEEAHGKLVLWSGLRNAPRCWAVIQPLLCAVYMPKCENDRVELPSRTLCQATRGPCA
RUO
SMO
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers