missing translation for 'onlineSavingsMsg'
Learn More

Human SNAP25 knockout SH-SY5Y cell lysate, 1 Kit

Product Code. 32140297 Shop All Abcam Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
32140297 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32140297 Supplier Abcam Supplier No. ab2801001Kit
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human SNAP25 knockout SH-SY5Y cell/tissue lysate.

  • Species: Human
  • Tissue: SNAP25 knockout SH-SY5Y

Specifications

For Use With (Application) Western Blot
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.