missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SPEG (aa 1257-1326) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53875 (PA5-53875. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q15772 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 10290 |
| Name | Human SPEG (aa 1257-1326) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | aortic preferentially expressed gene 1; aortic preferentially expressed protein 1; APEG1; APEG-1; BPEG; CNM5; KIAA1297; nuclear protein, marker for differentiated aortic smooth muscle and down-regulated with vascular injury; SPEG; SPEG complex locus; SPEGalpha; SPEGbeta; striated muscle preferentially expressed protein kinase |
| Common Name | SPEG |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction