Learn More
Abcam Human SS18 knockout HeLa cell line
Product Code. 32140444 Shop All Abcam Products
Change view
Click to view available options
Quantity:
2 x 1,000,000 Cells
Unit Size:
Pack of 2
Product Code. Quantity Unit Size
32140444 2 x 1,000,000 Cells Pack of 2
1 options
Product Code. 32140444 Supplier Abcam Supplier No. ab2654932x1000000Cells_
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
SS18 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp insertion in exon 4 and 2 bp insertion in exon 4. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To or...
SS18 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp insertion in exon 4 and 2 bp insertion in exon 4. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells