missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human STARD7 (aa 272-359) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66734 (PA5-66734. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May play a protective role in mucosal tissues by preventing exaggerated allergic responses. [UniProt]
Spezifikation
Spezifikation
| Accession Number | Q9NQZ5 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 56910 |
| Name | Human STARD7 (aa 272-359) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | AI852671; AL022671; AW544915; gestational trophoblastic tumor protein 1; GTT1; StAR related lipid transfer domain containing 7; StARD7; StAR-related lipid transfer (START) domain containing 7; StAR-related lipid transfer domain containing 7; stAR-related lipid transfer protein 7, mitochondrial; START domain containing 7; START domain-containing protein 7 |
| Common Name | STARD7 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur