missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Stem Cell Lineage Differentiation Package, mini, PeproTech®

Product Code. 30245173 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30245173 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30245173 Supplier Gibco™ Supplier No. HSCLDM
  • ONLINE EXCLUSIVE PRICE
    488.00 €
    351.36 € / Pack of 1
    Save 136.64 €
    28% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Human Stem Cell Lineage Differentiation Package, mini, contains the key stem cell differentiation factors required for the differentiation of stem cells into various cell lineages.

The Human Stem Cell Lineage Differentiation Package, mini, contains the key components required for the differentiation of stem cells into various cell lineages. The package contains the following three vials: 10 μg Human VEGF-165, 10 μg Human TGF-beta 1, and 10 μg Human/Mouse/Rat Activin A. VEGF-165 is a vascular endothelial growth factor that has been shown to be the fundamental regulator in the cell signaling of angiogenesis. TGF-beta 1 is a polypeptide member of the transforming growth factor beta superfamily of cytokines. Activins are dimeric growth and differentiation factors that belong to the transforming growth factor-beta (TGF-beta) superfamily of related signaling proteins.

VEGF-165 (Vascular Endothelial Growth Factor 165) is a key protein used in stem cell differentiation, particularly in the development of endothelial cells, which are essential for blood vessel formation (angiogenesis). VEGF-165 binds to its receptors on the surface of stem cells, activating signaling pathways that promote their differentiation into endothelial cells. By controlling stem cell differentiation with VEGF-165, researchers can improve the integration and functionality of transplanted tissues.

TGF-beta 1 is a polypeptide member of the transforming growth factor beta superfamily of cytokines. It helps regulate proliferation, cell differentiation, adhesion, and migration in many cell types. Many cells have TGF beta receptors, and the protein positively and negatively regulates many other growth factors. In the context of stem cell differentiation, TGF-beta 1is particularly important for inducing the differentiation of mesenchymal stem cells (MSCs) into chondrocytes, osteoblasts, and myofibroblasts.

Activins are dimeric growth and differentiation factors that belong to the transforming growth factor-beta (TGF-beta) superfamily of related signaling proteins. Activin A is a critical growth factor used in stem cell differentiation, primarily for directing the differentiation of pluripotent stem cells into definitive endoderm and mesoderm lineages. The precise control of stem cell differentiation using recombinant Activin A is essential for developing specific cell types.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Stem Cell Expansion Cytokine Package
Quantity 1 Ea.
Species Human
Format Lyophilized

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.