missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Stem Cell Renewal Package, mini, PeproTech®

Product Code. 30245175 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30245175 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30245175 Supplier Gibco™ Supplier No. HSCRM
  • ONLINE EXCLUSIVE PRICE
    231.00 €
    166.32 € / Pack of 1
    Save 64.68 €
    28% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Human Stem Cell Renewal Package, mini, contains the key stem cell growth factors required to maintain the self-renewal of your iPSCs in culture.

The Human Stem Cell Renewal Package, mini, contains the key stem cell growth factors required to maintain pluripotency and enable self-renewal of iPSCs in cell culture. The package contains the following two vials: 100 μg Human FGF-Basic (Cat. No. 100-18B-10UG) and 100 μg Human BMP-4 (Cat. No. 120-05ET-10UG). FGF-Basic (FGF2, FGFb, fibroblast growth factor) belongs to the fibroblast growth factor (FGF) family and interacts with high-affinity transmembrane receptors to influence cell proliferation and tissue neovascularization. BMP-4 is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily.

FGF-Basic (FGF2, FGFb, fibroblast growth factor) belongs to the fibroblast growth factor (FGF) family and interacts with high-affinity transmembrane receptors to influence cell proliferation and tissue neovascularization. FGF-Basic has been implicated in diverse biological processes, such as limb and nervous system development, wound healing, and tumor growth. FGF-basic plays a crucial role in maintaining the pluripotent state of iPSCs. It activates signaling pathways that prevent differentiation and promote self-renewal. FGF-basic also promotes cell proliferation, ensuring that iPSCs maintain a high growth rate.

BMP-4 is a member of the bone morphogenetic protein family, which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors.

TRUSTED_SUSTAINABILITY

Specifications

Format Lyophilized
Product Type Stem Cell Expansion Cytokine Package
Quantity 1 Ea.
Species Human

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.