missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human STIP1 Partial ORF (AAH02987.1, 80 a.a. - 180 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | AAH02987.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10963 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16055285
|
Abnova™
H00010963-Q03.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16045285
|
Abnova™
H00010963-Q03.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
STIP1 is an adaptor protein that coordinates the functions of HSP70 (see HSPA1A; MIM 140550) and HSP90 (see HSP90AA1; MIM 140571) in protein folding. It is thought to assist in the transfer of proteins from HSP70 to HSP90 by binding both HSP90 and substrate-bound HSP70. STIP1 also stimulates the ATPase activity of HSP70 and inhibits the ATPase activity of HSP90, suggesting that it regulates both the conformations and ATPase cycles of these chaperones (Song and Masison, 2005 [PubMed 16100115]).[supplied by OMIM]
Sequence: AALEFLNRFEEAKRTYEEGLKHEANNPQLKEGLQNMEARLAERKFMNPFNMPNLYQKLESDPRTRTLLSDPTYRELIEQLRNKPSDLGTKLQDPRIMTTLSSpecifications
AAH02987.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HOP/IEF-SSP-3521/P60/STI1/STI1L | |
STIP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10963 | |
STIP1 (Human) Recombinant Protein (Q03) | |
AALEFLNRFEEAKRTYEEGLKHEANNPQLKEGLQNMEARLAERKFMNPFNMPNLYQKLESDPRTRTLLSDPTYRELIEQLRNKPSDLGTKLQDPRIMTTLS | |
RUO | |
STIP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |