missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human STK29 Synthetic Peptide

Product Code. 15246689
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15246689 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15246689 Supplier Invitrogen™ Supplier No. PEP0478
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthetic Peptide

This peptide corresponds to 23 amino acids near the carboxy terminus of human BRSK2 PEP-0478 can be used as a blocking peptide with polyclonal antibody PA5-20358.

BRSK2 is a brain-selective serine/threonine kinase 2 that is mainly expressed in the brain, with weaker expression in testis and pancreas. BRSK2 expressed in insect cells specifically phosphorylates WEE1A, CDC25C and CDC25B in an in vitro assay. DNA damage induced by ultraviolet (UV) irradiation or methyl methane sulfonate, but not by ionizing radiation, enhanced endogenous BRSK2 kinase activity in a caffeine-sensitive manner and caused translocation of BRSK2 from the cytoplasm to the nucleus. Overexpression of BRSK2 in HeLa cells results in increased phosphorylation of CDC25C. DNA damage induced by ultraviolet (UV) irradiation or methyl methane sulfonate, but not by ionizing radiation, enhances endogenous BRSK2 kinase activity in a caffeine-sensitive manner and causes translocation of BRSK2 from the cytoplasm to the nucleus. Overexpression of BRSK2 induces G2/M arrest in HeLa cells. Small interfering RNA against BRSK2 partly abrogates UV-induced G2/M arrest. BRSK2 may act as a checkpoint kinase upon DNA damage induced by UV irradiation or methyl methane sulfonate.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 9024
Species Human
Purification Method purified
Accession Number Q8IWQ3
Common Name STK29
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 4833424K13Rik; BR serine/threonine kinase 2; BR serine/threonine-protein kinase 2; brain serine/threonine kinase 2; brain-selective kinase 2; Brain-specific serine/threonine-protein kinase 2; BRSK2; C11orf7; HUSSY-12; Kiaa4256; PEN11B; protein kinase SAD1B; SAD1; SAD1B; Sada; SAD-A; serine/threonine-protein kinase 29; Serine/threonine-protein kinase BRSK2; serine/threonine-protein kinase SAD-A; STK29; testicular tissue protein Li 30
Gene Symbol Brsk2
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.