missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Suspension Hepatocytes, Metabolism Qualified, Male, (4-8 x 106 cells)

Product Code. 30054510 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30054510 1 vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30054510 Supplier Gibco™ Supplier No. HMCS1S
  • ONLINE EXCLUSIVE PRICE
    962.00 €
    711.65 € / Pack of 1
    Save 250.35 €
    26% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human Suspension Hepatocytes, Metabolism Qualified, Male, (4-8 x 106 cells)

These cryopreserved human male hepatocytes are suitable for use in suspension metabolism applications.

Cells per vial: 4-8 million

The commonly tested Phase I (P450) and Phase II (UGT, SULT) drug metabolizing enzymes were analyzed for metabolic activity in each lot. Metabolite formation was measured by standard biochemical assays using LC⁄MS⁄MS or HPLC.

Get the Results You Need with GIBCO Hepatocytes
High quality hepatocytes, with high viabilities, in vivo-like enzyme expression levels, and proper cell morphology increase the ability to draw in vitro⁄in vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure you receive the highest quality cryopreserved hepatocytes.

Each lot is tested for:
  • Phase I and II enzyme activities
  • Proper morphology
  • Viabilities generally >75%

    For research use only. Not intended for human or animal therapeutic or diagnostic use.

    Related Links:

    Learn more about our cryopreserved hepatocytes

    Learn about our full line of hepatocytes ,

    Note:
    These products require shipment in LN2 vapor dewars therefore additional fees may apply.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Vials of cryopreserved hepatocytes should be stored in a cryostorage dewar or -135°C freezer.
    Cell Type Hepatocytes
    Culture Type Suspension Cell Culture
    Form Cryopreserved
    Gender Male
    Species Human
    Donor Source Single Donor
    No. of Cells 4-8 x 106
    Product Line Gibco
    Product Type Hepatocytes
    Quantity 1 vial
    Tested For Phase I and II Enzyme Activity, Proper Morphology, Viability Generally >75%
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.