missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Syndecan-3 (NP_055469) VersaClone cDNA
Description
- Also known as: SDCN; SYND3
- SDC3 belongs to the syndecan proteoglycan family.
- It may play a role in the organization of cell shape by affecting the actin cytoskeleton, possibly by transferring signals from the cell surface in a sugardependent mechanism.
- Allelic variants of SDC3 have been associated with obesity.
- The SDC3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hSDC3 |
| Accession Number | NP_055469 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1342 bp |
| Gene ID (Entrez) | 9672 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?