missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SYPL2 Synthetic Peptide

Product Code. 15349130
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15349130 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15349130 Supplier Invitrogen™ Supplier No. PEP0890

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

SYPL2 Synthetic Peptide, PEP-0890, from Invitrogen.

SYPL2, also known as Mitsugumin 29, was initially identified as a transmembrane protein from the triad junction in skeletal muscle that had significant homology with members of the synaptophysin family. SYPL2 is thought to participate in the excitation-contraction coupling process of skeletal muscle as SYPL2-null mice showed reduced muscle contractile force and altered triad junction structure and increased susceptibility to fatigue of the skeletal muscle. SYPL2 plays a critical role in muscle Ca2+ signaling by regulating the process of store-operated Ca2+ entry and interacts with ryanodine receptor (RyR), thereby influencing intracellular Ca2+ homeostasis through changes in the RyR/Ca2+ release function. Co-expression of SYPL2 and RyR in cultured cells leads to apoptotic cell death resulting from the depletion of Ca2+ from the intracellular stores.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 284612
Species Human
Purification Method purified
Accession Number Q5VXT5
Common Name Human SYPL2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol SYPL2
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.