Learn More
Abnova™ Human THBS4 Partial ORF (NP_003239.2, 401 a.a. - 500 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007060-Q01.10ug
Additional Details : Weight : 0.02000kg
Description
The protein encoded by this gene belongs to the thrombospondin protein family. Thrombospondin family members are adhesive glycoproteins that mediate cell-to-cell and cell-to-matrix interactions. This protein forms a pentamer and can bind to heparin and calcium. Studies of the rat counterpart suggest that this protein may be involved in local signaling in the developing and adult nervous system. [provided by RefSeq]
Sequence: YRCGPCKPGYTGDQIRGCKAERNCRNPELNPCSVNAQCIEERQGDVTCVCGVGWAGDGYICGKDVDIDSYPDEELPCSARNCKKDNCKYVPNSGQEDADRSpecifications
NP_003239.2 | |
Liquid | |
7060 | |
THBS4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TSP4 | |
THBS4 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
YRCGPCKPGYTGDQIRGCKAERNCRNPELNPCSVNAQCIEERQGDVTCVCGVGWAGDGYICGKDVDIDSYPDEELPCSARNCKKDNCKYVPNSGQEDADR | |
RUO | |
THBS4 | |
Wheat Germ (in vitro) | |
GST |