missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Thioredoxin-1 ELISA Kit

Product Code. 17135239
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17135239

Brand: Invitrogen™ EH451RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human Thioredoxin-1 quantitates human Thioredoxin-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Thioredoxin-1.

Thioredoxins (Trx) are small, multi-functional proteins with oxidoreductase activity and are ubiquitous in essentially all living cells. Trx contains a redox-active disulfide/dithiol group within the conserved Cys-Gly-Pro-Cys active site. The two cysteine residues in the conserved active centers can be oxidized to form intramolecular disulfide bonds. Reduction of the active site disulfide in oxidized Trx is catalyzed by Trx reductase with NADPH as the electron donor. The reduced Trx is a hydrogen donor for ribonucleotide reductase, the essential enzyme for DNA synthesis, and a potent general protein disulfide reductase with numerous functions in growth and redox regulations. Specific protein disulfide targets for reduction by Trx include protein disulfide-isomerase (PDI) and a number of transcription factors such as p53, NF-kB and AP-1 (T1-151). Trx is also capable of removing H2O2, particularly when it is coupled with either methionine sulfoxide reductase or several isoforms of peroxiredoxins.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number ADF; ATL-derived factor; AW550880; DKFZp686B1993; MGC61975; RP11-427L11.1; SASP; surface-associated sulphydryl protein; testicular tissue protein Li 199; thioredoxin; thioredoxin 1; thioredoxin delta 3; TRDX; TRX; TRX1; Txn; TXN delta 3; Txn1
Assay Range 0.2 to 50 ng/mL
Assay Sensitivity 0.2 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 7295
Gene Symbol TXN
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Thioredoxin 1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.