missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TIAM2 (aa 1562-1674) Control Fragment Recombinant Protein

Product Code. 30201524
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201524

Brand: Invitrogen™ RP90642

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (76%), Rat (76%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53098 (PA5-53098. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Modulates the activity of RHO-like proteins and connects extracellular signals to cytoskeletal activities. Acts as a GDP- dissociation stimulator protein that stimulates the GDP-GTP exchange activity of RHO-like GTPases and activates them. Mediates extracellular laminin signals to activate Rac1, contributing to neurite growth. Involved in lamellipodial formation and advancement of the growth cone of embryonic hippocampal neurons. Promotes migration of neurons in the cerebral cortex. When overexpressed, induces membrane ruffling accompanied by the accumulation of actin filaments along the altered plasma membrane. Activates specifically RAC1, but not CDC42 and RHOA.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8IVF5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 26230
Name Human TIAM2 (aa 1562-1674) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 3000002F19Rik; KIAA2016; mKIAA2016; RAS-related C3 botulinum substrate 1, guanine nucleotide exchange factor 1; SIF and TIAM1-like exchange factor; Stef; T cell lymphoma invasion and metastasis 2; T-cell lymphoma invasion and metastasis 2; T-cell lymphoma invasion and metastasis 2; RAS-related C3 botulinum substrate 1, guanine nucleotide exchange factor 1; TIAM2; TIAM-2; T-lymphoma invasion and metastasis-inducing protein 2
Common Name TIAM2
Gene Symbol Tiam2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IKESDILSDEDDDHRQTVKQGSPTKDIEIQFQRLRISEDPDVHPEAEQQPGPESGEGQKGGEQPKLVRGHFCPIKRKANSTKRDRGTLLKAQIRHQSLDSQSENATIDLNSVL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.