missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TMEFF2/Tomoregulin-2 (NP_057276.2) VersaClone cDNA

Product Code. 18784247
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18784247

Brand: R&D Systems™ RDC3244

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Gene TMEFF2/Tomoregulin-2
Accession Number NP_057276.2
Gene ID (Entrez) 23671
Gene Alias CT120.2, HPP1member 2, Hyperplastic polyposis protein 1, TENB2, Tomoregulin2, Tomoregulin-2, TPEFtomoregulin, TR-2, transmembrane protein TENB2, Transmembrane protein with EGF-like and two follistatin-like domains, transmembrane protein with EGF-like and two follistatin-like domains 2, TRtomoregulin-2
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.