missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TNF-alpha Quantikine ELISA Kit

Product Code. 30002173
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30002173 1 Kit 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30002173 Supplier R&D Systems™ Supplier No. DTA00D
  • ONLINE EXCLUSIVE PRICE
    854.30 €
    760.20 € / 96 tests
    Save 94.10 €
    11% Off
View all promotions
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human TNF-alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human TNF-alpha Quantikine ELISA Kit quantifies human TNF-alpha in cell culture supernates (50 ul), serum (50 ul), plasma (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 15.60 - 1000pg/mL
Assay Sensitivity 6.23pg/mL
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity HIV-1 Gag p24
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 7124
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Total Assay Time 4.0 hours
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.