missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Total IGF-I R/IGF1R DuoSet IC ELISA
Description
The Human Total IGF-I R/IGF1R DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human IGF-I R/IGF1R.
Specifications
Specifications
| Product Type | ELISA Development Kit for Intracellular Proteins |
| Quantity | 1 Kit |
| Species | Human |
| Conjugate | Biotin |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3480 |
| Synonym | CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF1R, IGF-1R, IGF-I receptor, IGFIR, IGF-IR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2 |
| Packaging Quantity | 5 Plates |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?