missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Total IGF-I R/IGF1R DuoSet IC ELISA

Product Code. p-4754550
Change view
Click to view available options
Packaging Quantity:
2 Plates
5 Plates
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity unitSize
30021884 5 Plates 1 set
30021187 2 Plates 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30021884 Supplier R&D Systems™ Supplier No. DYC3055

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Total IGF-I R/IGF1R DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Total IGF-I R/IGF1R DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human IGF-I R/IGF1R.
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Development Kit for Intracellular Proteins
Quantity 1 Kit
Species Human
Conjugate Biotin
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 3480
Synonym CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF1R, IGF-1R, IGF-I receptor, IGFIR, IGF-IR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2
Packaging Quantity 5 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.