missing translation for 'onlineSavingsMsg'
Learn More

Proteintech Human Total MMP-1 ELISA Kit

Product Code. 17224886 Shop All Proteintech Products
Change view
Click to view available options
Gene ID (Entrez):
4312
Unit Size:
96 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gene ID (Entrez) Unit Size
17224886 4312 96 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17224886 Supplier Proteintech Supplier No. KE0022396T
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Matrix metalloproteinase 1 (MMP1), which is also known as interstitial collagenase and fibroblast collagenase is a member of the matrix metalloproteinases (MMPs) family.

Proteins in this family mainly participate in the breakdown of extracellular matrix both in normal physiological processes and disease processes. Several factors contribute to the expression of MMP1, including endogenous factors, such as polymorphisms and epigenetic regulation in the promoter region of MMP1, as well as exogenous factors, such as the tumor microenvironment. A variety of MMP1 promoter genotypic polymorphisms have been reported in various ethnic populations, and their contributions to different disease risks have been explored. In the tumor microenvironment, a variety of inflammatory factors, including interleukin-8 (IL-8), IL-1β and tumor necrosis factor-α (TNF-α), have been reported to induce MMP1 expression in cancer cells. (PMID: 31115512, PMID: 28334049)

Specifications

Gene ID (Entrez) 4312
compliance-icons compliance-icons
Product Identifier
  • 96RXN Human Total MMP-1 ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.