missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TRIP12 (aa 1335-1428) Control Fragment Recombinant Protein

Product Code. 30200872
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30200872 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30200872 Supplier Invitrogen™ Supplier No. RP96152

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61116 (PA5-61116. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

E3 ubiquitin-protein ligase involved in ubiquitin fusion degradation (UFD) pathway and regulation of DNA repair. Part of the ubiquitin fusion degradation (UFD) pathway, a process that mediates ubiquitination of protein at their N-terminus, regardeless of the presence of lysine residues in target proteins. In normal cells, mediates ubiquitination and degradation of isoform p19ARF/ARF of CDKN2A, a lysine-less tumor suppressor required for p53/TP53 activation under oncogenic stress. In cancer cells, however, isoform p19ARF/ARF and TRIP12 are located in different cell compartments, preventing isoform p19ARF/ARF ubiquitination and degradation. Does not mediate ubiquitination of isoform p16-INK4a of CDKN2A. Also catalyzes ubiquitination of NAE1 and SMARCE1, leading to their degradation. Ubiquitination and degradation of target proteins is regulated by interaction with proteins such as MYC, TRADD or SMARCC1, which disrupt the interaction between TRIP12 and target proteins. Acts as a key regulator of DNA damage response by acting as a suppressor of RNF168, an E3 ubiquitin-protein ligase that promotes accumulation of 'Lys-63'-linked histone H2A and H2AX at DNA damage sites, thereby acting as a guard against excessive spreading of ubiquitinated chromatin at damaged chromosomes.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14669
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9320
Name Human TRIP12 (aa 1335-1428) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1110036I07Rik; 6720416K24Rik; AA410158; E3 ubiquitin-protein ligase for Arf; E3 ubiquitin-protein ligase TRIP12; gene trap locus 6; Gtl6; HECT-type E3 ubiquitin transferase TRIP12; KIAA0045; probable E3 ubiquitin-protein ligase TRIP12; thyroid hormone receptor interactor 12; thyroid receptor interacting protein 12; thyroid receptor interactor TRIP12; thyroid receptor-interacting protein 12; TR-interacting protein 12; TRIP12; TRIP-12; ULF
Common Name TRIP12
Gene Symbol TRIP12
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LNSGNVRHRLQFYIGEHLLPYNMTVYQAVRQFSIQAEDERESTDDESNPLGRAGIWTKTHTIWYKPVREDEESNKDCVGGKRGRAQTAPTKTSP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.