missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Human Uteroglobin ELISA Kit

Product Code. 16891506 Shop All Proteintech Products
Change view
Click to view available options
Target Species:
Human
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Target Species Unit Size
16891506 Human 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16891506 Supplier Proteintech™ Supplier No. KE0009996T
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

KE00099 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The Uteroglobin ELISA kit is to be used to detect and quantify protein levels of endogenous Uteroglobin.

  • The assay recognizes human Uteroglobin. An antibody specific for Uteroglobin has been pre-coated onto the microwells.
  • The Uteroglobin protein in samples is captured by the coated antibody after incubation. Following extensive washing, another antibody specific for Uteroglobin is added to detect the captured Uteroglobin protein.
  • For signal development, horseradish peroxidase (HRP)-conjugated antibody is added, followed by Tetramethyl-benzidine (TMB) reagent. Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.
  • Uteroglobin, also named clara cell secretory protein or CC16, is mainly secreted by the club cells along the tracheobronchial tree, diffuses passively into the bloodstream and is rapidly eliminated by glomerular filtration.
  • It is a steroid-dependent, immunomodulatory, cytokine-like protein, and it is secreted by the mucosal epithelial cells of many vertebrates and takes part in some signaling pathways.
  • Growing evidence indicates that Uteroglobin has anti-inflammatory and anti-toxicant properties in the lung and may protect against obstructive lung diseases.
  • Lower levels of Uteroglobin in blood and airways have been associated with prevalence and severity of Chronic obstructive pulmonary disease (COPD). Defects in Uteroglobin are associated with a susceptibility to asthma.

Specifications

Accession Number NP_003348
Assay Range 0.156 to 10 ng/mL
Assay Sensitivity 0.03 ng/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Human
Gene ID (Entrez) 7356
Host Species Human
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 4.5 hr.
UniProt ID P11684
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human Uteroglobin ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.