missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human V-ATPase C1 (aa 187-267) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66075 (PA5-66075. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
ATP6V1C1 is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of intracellular compartments of eukaryotic cells. V-ATPase dependent acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This gene is one of two genes that encode the V1 domain C subunit proteins and is found ubiquitously. This C subunit is analogous but not homologous to gamma subunit of F-ATPases. Previously, this gene was designated ATP6D.
Specifications
Specifications
| Accession Number | P21283 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 528 |
| Name | Human V-ATPase C1 (aa 187-267) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1700025B18Rik; ATP6C; Atp6c1; ATP6D; Atp6v1c1; ATPase H+ transporting V1 subunit C1; ATPase, H+ transporting, lysosomal 42 kDa, V1 subunit C1; ATPase, H+ transporting, lysosomal V1 subunit C1; ATPase, H+ transporting, V1 subunit C, isoform 1; FLJ20057; H(+)-transporting two-sector ATPase, subunit C; H+ -ATPase C subunit; H+-transporting ATPase chain C, vacuolar; subunit C of vacuolar proton-ATPase V1 domain; testicular tissue protein Li 223; U13839; vacuolar ATP synthase subunit C; vacuolar H+ -ATPase C subunit; vacuolar proton pump C subunit; vacuolar proton pump subunit C 1; vacuolar proton pump, 42-kD subunit; vacuolar proton-ATPase, subunit C, VI domain; vatC; V-ATPase C subunit; V-ATPase subunit C 1; Vma5; V-type proton ATPase subunit C 1 |
| Common Name | V-ATPase C1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction