missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human VDAC1 Partial ORF (AAH08482.1, 64 a.a. - 145 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007416-Q02.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16044945

  • 508.00 € / 25µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: WTEYGLTFTEKWNTDNTLGTEITVEDQLARGLKLTFDSSFSPNTGKKNAKIKTGYKREHINLGCDMDFDIAGPSIRGALVLG
Specifications

Specifications

AAH08482.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.65kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
WTEYGLTFTEKWNTDNTLGTEITVEDQLARGLKLTFDSSFSPNTGKKNAKIKTGYKREHINLGCDMDFDIAGPSIRGALVLG
RUO
VDAC1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
7416
VDAC1 (Human) Recombinant Protein (Q02)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC111064/PORIN/PORIN-31-HL
VDAC1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers