missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human VEGF Quantikine ELISA Kit

Product Code. 18712054
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18712054 1 Plate 1 set
18732114 6 Plates 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18712054 Supplier R&D Systems™ Supplier No. DVE00
  • ONLINE EXCLUSIVE PRICE
    854.30 €
    760.20 € / 1 sets
    Save 94.10 €
    11% Off
View all promotions
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human VEGF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human VEGF Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human VEGF in cell culture supernates (200 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 15.60 to 1000 pg/mL (Cell Culture Supernates), 31.30 to 2000 pg/mL (Serum, Heparin Plasma, Citrate Plasma, EDTA Plasma)
Assay Sensitivity 5 pg/mL (Cell Culture Supernates), 9 pg/mL (Serum, Heparin Plasma, Citrate Plasma, EDTA Plasma)
Conjugate HRP
Cross Reactivity Cross-species reactivity observed with 1 or more species tested. <0.5% cross-reactivity observed with available related molecules.
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Specificity Natural and Recombinant Human VEGF. This assay also recognizes Recombinant Human VEGF165b.
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 7422
Quantity 1 Plate
Sample Volume Cell Culture Supernates (200uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL),Citrate Plasma (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.