missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human VEGFR2/KDR Quantikine ELISA Kit

Product Code. p-4753976
Click to view available options
Quantity:
1 Plate
6 Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18792064

Brand: R&D Systems™ DVR200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human VEGFR2/KDR Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human VEGFR2/KDR Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human VEGFR2/KDR/Flk-1 in cell culture supernates (100 ul), cell lysates (100 ul), serum (20 ul), edta plasma (20 ul), heparin plasma (20 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 78.10 - 5000pg/mL
Assay Sensitivity 11.4pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum
Specificity Natural and Recombinant Human VEGF R2
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3791
Quantity 1 Plate
Sample Volume Cell Culture Supernates (100uL),Cell Lysates (100uL),Serum (20uL),EDTA Plasma (20uL),Heparin Plasma (20uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, Flk1, Flk-1, FLK1tyrosine kinase growth factor receptor, KDR, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), KRD1, Ly73, Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGF R2, VEGFR, VEGFR2, VEGFR-2
Target VEGFR2/KDR/Flk-1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.