missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human WDR97 (aa 1293-1382) Control Fragment Recombinant Protein

Product Code. 30208233
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30208233

Brand: Invitrogen™ RP101969

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (48%), Rat (48%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84416 (PA5-84416. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The function and protein-coding potential of this gene is unknown. The exon combination is based on AB058778.1 for which two possible open reading frames can be predicted (with start codons at nucleotide 26 or 2614). The position of the first ORF stop codon is consistent with a prediction of nonsense-mediated decay. Given the observation of the first ORF and its length, a predicted translation of the second ORF is inconsistent with the translation leaky scanning theory. Therefore, this gene is represented as a non-protein-coding transcript. [provided by RefSeq, Oct 2008].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number A6NE52
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 340390
Name Human WDR97 (aa 1293-1382) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias KIAA1875; WD repeat domain 97; WD repeat-containing protein 97; WDR97
Common Name WDR97
Gene Symbol WDR97
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LMSYFLYSPVHCRPELKKLLHGLGLQDPEGFLFKEMMTWVQGPDLDSKAGLRTCCHQKLEDMIQELQETPSQTSVVSGAPTRASVIPSGT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.