missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZDHHC16 (aa 221-265) Control Fragment Recombinant Protein

Product Code. 30207026
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30207026 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30207026 Supplier Invitrogen™ Supplier No. RP104716

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (62%), Rat (62%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65260 (PA5-65260. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ZDHHC16 (zinc finger, DHHC-type containing 16), also known as APH2, is a 377 amino acid multi-pass membrane protein that localizes to the endoplasmic reticulum and contains one DHHC-type zinc finger. Existing as multiple alternatively spliced isoforms, ZDHHC16 interacts with c-Abl and catalyzes the conversion of Palmitoyl-CoA and protein-cysteine to S-palmitoyl protein and CoA. Via its association with c-Abl, ZDHHC16 may be involved in the regulation of apoptosis. The gene encoding ZDHHC16 maps to human chromosome 10, which houses over 1,200 genes and comprises nearly 4.5% of the human genome. Defects in some of the genes that map to chromosome 10 are associated with Charcot-Marie Tooth disease, Jackson-Weiss syndrome, Usher syndrome, nonsyndromatic deafness, Wolman's syndrome, Cowden syndrome, multiple endocrine neoplasia type 2 and porphyria.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q969W1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84287
Name Human ZDHHC16 (aa 221-265) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1500015N03Rik; Abl-philin 2; ablphilin-2; Aph2; DHHC-16; membrane-associated DHHC16 zinc finger protein; Palmitoyltransferase ZDHHC16; probable palmitoyltransferase ZDHHC16; UNQ2570/PRO6258; Zdhhc16; Zinc finger DHHC domain-containing protein 16; zinc finger DHHC-type containing 16; zinc finger, DHHC domain containing 16; zinc finger, DHHC-type containing 16
Common Name ZDHHC16
Gene Symbol ZDHHC16
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LFREAYAAIEKMKQLDKNKLQAVANQTYHQTPPPTFSFRERMTHK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.