missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZIC4 Partial ORF (NP_115529, 1 a.a. - 90 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16035715
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16035715 10 μg 10µg
16045715 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16035715 Supplier Abnova™ Supplier No. H00084107Q02.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. Members of this family are important during development, and have been associated with X-linked visceral heterotaxy and holoprosencephaly type 5. This gene is closely linked to the gene encoding zinc finger protein of the cerebellum 1, a related family member on chromosome 3. This gene encodes a protein of unknown function. [provided by RefSeq]

Sequence: MRYKTSLVMRKRLRLYRNTLKESSSSSGHHGPQLTAASSPSVFPGLHEEPPQASPSRPLNGLLRLGLPGDMYARPEPFPPGPAARSDALA

Specifications

Accession Number NP_115529
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 84107
Molecular Weight (g/mol) 35.64kDa
Name ZIC4 (Human) Recombinant Protein (Q02)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Immunogen MRYKTSLVMRKRLRLYRNTLKESSSSSGHHGPQLTAASSPSVFPGLHEEPPQASPSRPLNGLLRLGLPGDMYARPEPFPPGPAARSDALA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ42609/FLJ45833
Common Name ZIC4
Gene Symbol ZIC4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.