missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZMYM2 Synthetic Peptide

Product Code. 15215209
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15215209 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15215209 Supplier Invitrogen™ Supplier No. PEP1204
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Synthetic Peptide

This peptide corresponds to 17 amino acids near the amino terminus of human ZMYM2. PEP-1204 can be used as a blocking peptide with polyclonal antibody PA5-21090.

ZNF198 is a ubiquitously expressed nuclear protein whose physiological role is largely unknown. It consists of 5 N-terminal zinc finger motifs responsible for protein-protein interactions, a central proline-rich domain and C-terminal nuclear localization signal (NLS) and an acidic domain. Reciprocal chromosomal translocation involving ZNF198 and FGFR1 results in the formation of ZNF198/FGFR1 fusion kinase protein. This fusion protein is a ligand-dependent, constitutively active cytoplasmic tyrosine kinase and regulates several STAT transcription factors including STAT 1, 3 and 5. This fusion protein is an oncogenic protein and is associated with an atypical myeloproliferative disease, peripheral blood eosinophilia and T-cell leukemia/lymphoma.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 7750
Species Human
Purification Method purified
Accession Number Q9UBW7
Common Name ZMYM2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 5830413P05Rik; FIM; Fused in myeloproliferative disorders protein; MYM; RAMP; rearranged in an atypical myeloproliferative disorder; Rearranged in atypical myeloproliferative disorder protein; RP11-264J4.7; SCLL; Zfp198; zinc finger MYM-type containing 2; zinc finger MYM-type protein 2; Zinc finger protein 198; zinc finger, MYM-type 2; Zmym2; ZNF198
Gene Symbol ZMYM2
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.