missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ HumanKine™ Recombinant Human BMP-7 Protein
Shop All Proteintech Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Description
- Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Specifications
Specifications
| Gene ID (Entrez) | 655 |
| Molecular Weight (g/mol) | 29 kDa, homodimer, glycosylated |
| Quantity | 10 μg |
| Species | Human |
| Reconstitution | Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in 10 mM acetic acid containing 0.1% endotoxin-free recombinant human serum albumin (HSA). |
| Content And Storage | Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. |
| Activity | Typically ≤ 100 ng/mL EC50 |
| Expression System | Human embryonic kidney 293 (HEK293) human cell expression system |
| Form | Lyophilized |
| Purity or Quality Grade | > 95% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction