Learn More
Proteintech™ HumanKine™ Recombinant Human FGF Basic-TS Protein
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
Brand: Proteintech™ HZ-1285-10UG
This item is not returnable.
View return policy
Description
- Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Specifications
| 2247 | |
| 10 μg | |
| Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA). | |
| Typically ≤ 0.5 ng/mL EC50 | |
| Lyophilized |
| 17 kDa, monomer, non-glycosylated | |
| Human | |
| Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. | |
| Human embryonic kidney 293 (HEK293) human cell expression system | |
| > 95% |