missing translation for 'onlineSavingsMsg'
Learn More

Humic acid, sodium salt, 45-70%, technical, (as humic acid)

Product Code. 10102920 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
1kg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10102920 100 g Plastic bottle 100g
10284820 5 g Glass bottle 5g
10560211 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10102920 Supplier Thermo Scientific Acros Supplier No. 120861000
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 68131-04-4 | C9H8Na2O4 | 226.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68131-04-4
Molecular Formula C9H8Na2O4
Molecular Weight (g/mol) 226.139
MDL Number MFCD00135560
InChI Key HMGUIQPKFUZDPV-UHFFFAOYSA-L
Synonym humic acid sodium salt, sodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, sodium humate, disodium bicyclo 2.2.1 hept-5-en-2,3-dicarboxylate, disodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, bicyclo 2.2.1 hepta-5-ene-2,3-dicarboxylic acid disodium salt
PubChem CID 21908873
IUPAC Name disodium;bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate
SMILES C1C2C=CC1C(C2C(=O)[O-])C(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >300.0°C
Color Black
Quantity 100 g
Infrared Spectrum Authentic
Loss on Drying 14% to 25% (105°C, 4 hrs)
Packaging Plastic bottle
Physical Form Crystalline Powder
Assay Percent Range 45-70%
Percent Purity 45 to 70%
Grade Technical
Chemical Name or Material Humic acid, sodium salt, tech., 50-60% (as humic acid)
Show More Show Less
compliance-icons
Product Identifier
  • Humic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Humic acids, sodiumsalts

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.