missing translation for 'onlineSavingsMsg'
Learn More

Humic acid, sodium salt, 45-70%, technical, (as humic acid)

Product Code. 10284820 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
1kg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10284820 5 g Glass bottle 5g
10102920 100 g Plastic bottle 100g
10560211 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10284820 Supplier Thermo Scientific Acros Supplier No. 120860050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 68131-04-4 | C9H8Na2O4 | 226.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68131-04-4
Molecular Formula C9H8Na2O4
Molecular Weight (g/mol) 226.139
MDL Number MFCD00135560
InChI Key HMGUIQPKFUZDPV-UHFFFAOYSA-L
Synonym humic acid sodium salt, sodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, sodium humate, disodium bicyclo 2.2.1 hept-5-en-2,3-dicarboxylate, disodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, bicyclo 2.2.1 hepta-5-ene-2,3-dicarboxylic acid disodium salt
PubChem CID 21908873
IUPAC Name disodium;bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate
SMILES C1C2C=CC1C(C2C(=O)[O-])C(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >300.0°C
Color Black
Assay Percent Range 45-70%
Percent Purity 45 to 70%
Grade Technical
Quantity 5 g
Infrared Spectrum Authentic
Loss on Drying 14% to 25% (105°C, 4 hrs)
Packaging Glass bottle
Physical Form Crystalline Powder
Chemical Name or Material Humic acid, sodium salt, tech., 50-60% (as humic acid)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.