Learn More
Hamilton™ HV 4-2 Valve, 4 ports, 90 deg Flow Path, aluminum housing
Product Code. 11570052
Change view
Click to view available options
Manual valve
Product Code.
11570052 Manual valve
1 options
Product Code. 11570052 Supplier Hamilton™ Supplier No. 86731
  • ONLINE EXCLUSIVE PRICE
    171.00 €
    153.31 € / Pack of 1
    Save 17.69 €
    10% Off
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Providing an economical and flexible method of custom-building a fluid flow control system.
  • Chemically inert flow path
  • Ideally suited for low pressure applications
  • Leak-tight seals