Learn More
Hamilton™ HVC Standard Ceramic Valves
Product Code. 11543314
Click to view available options
Material:
Aluminum
Stainless Steel
Model:
3-2 Model
3-5 Model
T 3-2 Model
Unit Size:
Each
Product Code. 11543314
Brand: Hamilton™ 520801
Designed to provide superior performance with salt solutions.
  • Rigid ceramic fluid path is resistant to scratching from salt crystals
  • Provides superior life and performance
  • Exceptionally easy to service and modify
  • Maximizes flexibility and value
  • Ideal for low pressure applications
  • All HVC valves have a thread depth of 1/4 in.-28 UNF
Specifications
Description 3-2 Model, 3 Ports, Aluminum Housing
For Use With PSD/2 and 3 Syringe Drive Modules
Material Aluminum
Model 3-2 Model
Type Ceramic HV Valve