missing translation for 'onlineSavingsMsg'
Learn More

Hyaluronic acid, MedChemExpress

Product Code. 16498396
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
1g
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498396 100 mg 100mg
16494427 50 mg 50mg
16462186 500 mg 500mg
16441436 200 mg 200mg
16452249 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16498396 Supplier Medchem Express Supplier No. HYB0633A
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Hyaluronic acid (corn fermented) is a biopolymer composed of repeating units of disaccharides with various applications.

Hyaluronic acid (corn fermented) is a biopolymer composed of repeating units of disaccharides with various applications.

Specifications

Chemical Name or Material Hyaluronic acid
Color White
CAS 9004-61-9
Physical Form Solid
Purity Grade Notes Research
Molecular Formula (C14H21NO11)n
Synonym Hyaluronate
Solubility Information H2O : 14.29 mg/mL (33.12 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.32 mM; ultrasonic and warming and heat to 80°C)
Quantity 100 mg
SMILES CCC1[C@@H](O)[C@@H](O)O[C@@H](O[C@H]2[C@H](C(O)=O)OC(OC)[C@@H](O)[C@@H]2O)[C@@H]1NC(C)=O.[n]
Molecular Weight (g/mol) 431.49
For Use With (Application) Cancer-programmed cell death
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.